Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025010.4 | NP_079286.2 | 2 - 3 | 389 | 67 |
XM_005265001.3 | XP_005265058.2 | 2 - 3 | 289 | 67 | |
XM_005265002.4 | XP_005265059.2 | 2 - 3 | 289 | 67 | |
XM_005265003.3 | XP_005265060.2 | 2 - 3 | 289 | 67 | |
XM_006713079.1 | XP_006713142.1 | 2 - 3 | 245 | 67 | |
GenBank mRNA | AB062478.1 | - | 2 - 3 | 561 | 67 |
AI356423.1 | - | 2 - 3 | 389 | 67 | |
AK023765.1 | - | 2 - 3 | 283 | 67 | |
AK291477.1 | - | 2 - 3 | 283 | 67 | |
BC032620.1 | - | 2 - 3 | 201 | 67 | |
BX537953.1 | - | 2 - 3 | 280 | 67 |