Assay Design



Gene Transcripts

Gene Symbol
CCDC68
Entrez Gene ID
80323
Gene Name
coiled-coil domain containing 68
Gene Aliases
SE57-1
Chromosome Location
Chr.18: 54901509 - 54959539 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143829.1 NP_001137301.1 4 - 5 381 95
NM_025214.2 NP_079490.1 4 - 5 484 95
XM_006722552.3 XP_006722615.1 4 - 5 433 95
XM_011526198.2 XP_011524500.1 4 - 5 1493 95
XM_011526199.2 XP_011524501.1 4 - 5 392 95
XM_011526200.2 XP_011524502.1 4 - 5 545 95
XM_011526201.2 XP_011524503.1 4 - 5 1452 95
XM_011526202.1 XP_011524504.1 4 - 5 484 95
XM_011526204.2 XP_011524506.1 4 - 5 484 95
GenBank mRNA AF273051.1 - 4 - 5 382 95
AK058097.1 - 4 - 5 381 95
AK313793.1 - 2 - 3 227 95
BC029508.1 - 4 - 5 438 95

View Full Product Details