Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030764.3 | NP_110391.2 | 10 - 11 | 1517 | 115 |
XM_011509974.2 | XP_011508276.1 | 10 - 11 | 1852 | 141 | |
GenBank mRNA | AF319438.1 | - | 5 - 6 | 1002 | 115 |
AF390037.1 | - | 5 - 6 | 840 | 115 | |
AF459633.1 | - | 10 - 11 | 1505 | 115 | |
AL833194.1 | - | 5 - 6 | 3376 | 115 | |
AY043465.1 | - | 10 - 11 | 1504 | 115 | |
AY358130.1 | - | 10 - 11 | 1507 | 115 | |
BC129836.1 | - | 5 - 6 | 842 | 115 |