Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030787.3 | NP_110414.1 | 9 - 10 | 1646 | 110 |
XM_011510020.2 | XP_011508322.1 | 9 - 10 | 1553 | 110 | |
GenBank mRNA | AF295327.1 | - | 9 - 10 | 1611 | 110 |
AK314026.1 | - | 9 - 10 | 1647 | 110 | |
BC111773.1 | - | 9 - 10 | 1611 | 110 |