Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030816.4 | NP_110443.3 | 9 - 10 | 1543 | 147 |
XM_005271234.3 | XP_005271291.1 | 9 - 10 | 1786 | 147 | |
XM_006710929.3 | XP_006710992.1 | 9 - 10 | 1556 | 147 | |
XM_017002413.1 | XP_016857902.1 | 7 - 8 | 766 | 147 | |
XM_017002414.1 | XP_016857903.1 | 8 - 9 | 848 | 147 | |
XM_017002415.1 | XP_016857904.1 | 8 - 9 | 827 | 147 | |
XM_017002416.1 | XP_016857905.1 | 4 - 5 | 512 | 147 | |
GenBank mRNA | AK074187.1 | - | 1 - 2 | 167 | 147 |
AL136717.1 | - | 8 - 9 | 1439 | 147 | |
BC028840.1 | - | 9 - 10 | 1532 | 147 |