Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001973.3 | NP_001964.2 | 3 - 4 | 2337 | 111 |
XM_005244950.4 | XP_005245007.1 | 4 - 5 | 1247 | 111 | |
XM_005244951.4 | XP_005245008.1 | 3 - 4 | 2041 | 111 | |
XM_017000580.1 | XP_016856069.1 | 3 - 4 | 2491 | 111 | |
GenBank mRNA | BC040145.1 | - | 3 - 4 | 1358 | 111 |
M85165.1 | - | 3 - 4 | 1236 | 111 |