Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271043.2 | NP_001257972.1 | 7 - 8 | 1156 | 94 |
NM_001271044.2 | NP_001257973.1 | 7 - 8 | 1116 | 94 | |
NM_002501.3 | NP_002492.2 | 7 - 8 | 1147 | 94 | |
XM_005259918.4 | XP_005259975.1 | 7 - 8 | 1154 | 94 | |
XM_005259919.3 | XP_005259976.1 | 7 - 8 | 1271 | 94 | |
XM_005259920.3 | XP_005259977.1 | 7 - 8 | 1833 | 94 | |
XM_011528040.1 | XP_011526342.1 | 7 - 8 | 1154 | 94 | |
GenBank mRNA | AK295290.1 | - | 7 - 8 | 1156 | 94 |
AK297261.1 | - | - | 1115 | 94 | |
BC117113.1 | - | 6 - 7 | 1110 | 94 | |
BC117115.1 | - | 6 - 7 | 1110 | 94 | |
FJ861276.1 | - | 6 - 7 | 1055 | 94 | |
L31881.1 | - | 6 - 7 | 1115 | 94 | |
U18759.1 | - | 7 - 8 | 1096 | 94 | |
U18760.1 | - | 6 - 7 | 737 | 94 | |
U18761.1 | - | 6 - 7 | 1162 | 94 |