Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002660.2 | NP_002651.2 | 1 - 2 | 342 | 84 |
NM_182811.1 | NP_877963.1 | 1 - 2 | 342 | 84 | |
XM_005260438.2 | XP_005260495.1 | 1 - 2 | 344 | 84 | |
XM_011528865.1 | XP_011527167.1 | 1 - 2 | 131 | 84 | |
XM_011528867.2 | XP_011527169.1 | 1 - 2 | 342 | 84 | |
GenBank mRNA | AB210028.1 | - | 1 - 2 | 626 | 84 |
BC136466.1 | - | 1 - 2 | 309 | 84 | |
BC144136.1 | - | 1 - 2 | 309 | 84 | |
BX415739.2 | - | 1 - 2 | 349 | 84 | |
M34667.1 | - | 1 - 2 | 297 | 84 |