Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003239.3 | NP_003230.1 | 1 - 2 | 1482 | 76 |
GenBank mRNA | AK313564.1 | - | 1 - 2 | 563 | 76 |
BC018503.1 | - | 1 - 2 | 453 | 76 | |
BT007287.1 | - | 1 - 2 | 353 | 76 | |
EF560714.1 | - | 1 - 2 | 466 | 76 | |
J03241.1 | - | 1 - 2 | 615 | 76 | |
X14149.1 | - | 1 - 2 | 606 | 76 |