Assay Design
Gene Transcripts
- Gene Symbol
- DEFA1
- Entrez Gene ID
-
1667
- Gene Name
- defensin alpha 1
- Gene Aliases
-
DEF1, DEFA2, HNP-1, HP-1, HP1, MRS
- Chromosome Location
-
Chr.8: 6977649 - 6980119 on Build GRCh38
- Gene Symbol
- DEFA1B
- Entrez Gene ID
-
728358
- Gene Name
- defensin alpha 1B
- Gene Aliases
-
HNP-1, HP-1, HP1
- Chromosome Location
-
Chr.8: 6996766 - 6999202 on Build GRCh38
- Gene Symbol
- DEFA3
- Entrez Gene ID
-
1668
- Gene Name
- defensin alpha 3
- Gene Aliases
-
DEF3, HNP-3, HNP3, HP-3, HP3
- Chromosome Location
-
Chr.8: 7015869 - 7018336 on Build GRCh38
View Full Product Details