Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000028.2 | NP_000019.2 | 24 - 25 | 3739 | 73 |
NM_000642.2 | NP_000633.2 | 24 - 25 | 3661 | 73 | |
NM_000643.2 | NP_000634.2 | 24 - 25 | 3459 | 73 | |
NM_000644.2 | NP_000635.2 | 24 - 25 | 3399 | 73 | |
NM_000645.2 | NP_000636.2 | 22 - 23 | 3472 | 73 | |
NM_000646.2 | NP_000637.2 | 24 - 25 | 3472 | 73 | |
XM_005270557.2 | XP_005270614.1 | 24 - 25 | 3354 | 73 | |
XM_017000501.1 | XP_016855990.1 | 12 - 13 | 1588 | 73 | |
GenBank mRNA | AB208867.1 | - | 23 - 24 | 3265 | 73 |
AB621804.1 | - | 24 - 25 | 3609 | 73 | |
BC078663.1 | - | 24 - 25 | 3381 | 73 | |
M85168.1 | - | 22 - 23 | 3474 | 73 | |
U84007.1 | - | 24 - 25 | 3661 | 73 | |
U84008.1 | - | 24 - 25 | 3399 | 73 | |
U84009.1 | - | 24 - 25 | 3459 | 73 | |
U84010.1 | - | 24 - 25 | 3739 | 73 | |
U84011.1 | - | 24 - 25 | 3472 | 73 |