Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000899.4 | NP_000890.1 | 2 - 3 | 329 | 123 |
NM_003994.5 | NP_003985.2 | 2 - 3 | 329 | 123 | |
GenBank mRNA | AF119835.1 | - | 2 - 3 | 276 | 123 |
AF400436.1 | - | 2 - 3 | 317 | 123 | |
AF400437.1 | - | 2 - 3 | 276 | 123 | |
AK290319.1 | - | 2 - 3 | 325 | 123 | |
AK293002.1 | - | 2 - 3 | 245 | 123 | |
BC069733.1 | - | 2 - 3 | 290 | 123 | |
BC069783.1 | - | 2 - 3 | 291 | 123 | |
BC069797.1 | - | 2 - 3 | 286 | 123 | |
BC074725.2 | - | 2 - 3 | 174 | 123 | |
BC126166.1 | - | 2 - 3 | 284 | 123 | |
BC143899.1 | - | 2 - 3 | 284 | 123 | |
DC320486.1 | - | 2 - 3 | 329 | 123 | |
M59964.1 | - | 2 - 3 | 317 | 123 |