Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258462.1 | NP_001245391.1 | 5 - 6 | 608 | 149 |
NM_001258463.1 | NP_001245392.1 | 5 - 6 | 546 | 149 | |
NM_001310158.1 | NP_001297087.1 | 5 - 6 | 649 | 149 | |
NM_001310160.1 | NP_001297089.1 | 1 - 2 | 1532 | 149 | |
NM_001310161.1 | NP_001297090.1 | 2 - 3 | 415 | 149 | |
NM_001604.5 | NP_001595.2 | 5 - 6 | 596 | 149 | |
GenBank mRNA | AB593092.1 | - | 5 - 6 | 547 | 149 |
AB593093.1 | - | 5 - 6 | 546 | 149 | |
AB593094.1 | - | 5 - 6 | 649 | 149 | |
AK094249.1 | - | 1 - 2 | 1342 | 149 | |
BX640762.1 | - | 5 - 6 | 575 | 149 | |
DA089215.1 | - | 2 - 3 | 415 | 149 | |
DA141443.1 | - | 5 - 6 | 514 | 149 | |
DA571138.1 | - | 2 - 3 | 237 | 149 | |
KP255960.1 | - | 1 - 2 | 139 | 149 |