Assay Design



Gene Transcripts

Gene Symbol
FXYD2
Entrez Gene ID
486
Gene Name
FXYD domain containing ion transport regulator 2
Gene Aliases
ATP1G1, HOMG2
Chromosome Location
Chr.11: 117820075 - 117828092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001680.4 NP_001671.2 1 - 2 96 108
GenBank mRNA AF241236.1 - 1 - 2 71 108
BC013289.1 - 1 - 2 99 108
U50743.1 - 1 - 2 106

View Full Product Details