Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204426.1 | NP_001191355.1 | 6 - 7 | 878 | 97 |
NM_002314.3 | NP_002305.1 | 7 - 8 | 1100 | 97 | |
GenBank mRNA | AB209557.1 | - | 4 - 5 | 572 | 97 |
AK026544.1 | - | 1 - 2 | 136 | 97 | |
AK290162.1 | - | 8 - 9 | 1025 | 97 | |
AK300382.1 | - | 6 - 7 | 817 | 97 | |
D26309.1 | - | 8 - 9 | 981 | 97 |