Assay Design



Gene Transcripts

Gene Symbol
MBD1
Entrez Gene ID
4152
Gene Name
methyl-CpG binding domain protein 1
Gene Aliases
CXXC3, PCM1, RFT
Chromosome Location
Chr.18: 50266882 - 50281774 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204136.1 NP_001191065.1 2 - 3 549 61
NM_001204137.1 NP_001191066.1 2 - 3 549 61
NM_001204138.1 NP_001191067.1 2 - 3 549 61
NM_001204139.1 NP_001191068.1 2 - 3 549 61
NM_001204140.1 NP_001191069.1 2 - 3 549 61
NM_001204141.1 NP_001191070.1 2 - 3 549 61
NM_001204142.1 NP_001191071.1 2 - 3 549 61
NM_001204143.1 NP_001191072.1 2 - 3 330 61
NM_001204151.2 NP_001191080.1 2 - 3 549 61
NM_001323942.1 NP_001310871.1 2 - 3 549 61
NM_001323947.1 NP_001310876.1 2 - 3 549 61
NM_001323950.1 NP_001310879.1 2 - 3 549 61
NM_001323951.1 NP_001310880.1 2 - 3 549 61
NM_001323952.1 NP_001310881.1 2 - 3 549 61
NM_001323954.1 NP_001310883.1 2 - 3 549 61
NM_002384.2 NP_002375.1 2 - 3 549 61
NM_015844.2 NP_056669.2 2 - 3 549 61
NM_015845.3 NP_056670.2 2 - 3 549 61
NM_015846.3 NP_056671.2 2 - 3 549 61
NM_015847.3 NP_056723.2 2 - 3 549 61
XM_005258271.2 XP_005258328.1 2 - 3 296 61
XM_006722456.2 XP_006722519.1 2 - 3 296 61
XM_011525991.1 XP_011524293.1 2 - 3 296 61
XM_011525993.2 XP_011524295.1 2 - 3 296 61
XM_011525994.2 XP_011524296.1 2 - 3 296 61
XM_011525998.2 XP_011524300.1 2 - 3 296 61
XM_011525999.1 XP_011524301.1 2 - 3 296 61
XM_011526001.1 XP_011524303.1 2 - 3 296 61
XM_011526002.1 XP_011524304.1 2 - 3 296 61
XM_011526003.2 XP_011524305.1 2 - 3 296 61
XM_011526006.1 XP_011524308.1 2 - 3 296 61
XM_011526007.1 XP_011524309.1 2 - 3 296 61
XM_017025751.1 XP_016881240.1 2 - 3 296 61
XM_017025752.1 XP_016881241.1 2 - 3 296 61
XM_017025753.1 XP_016881242.1 2 - 3 296 61
XM_017025754.1 XP_016881243.1 2 - 3 296 61
XM_017025755.1 XP_016881244.1 2 - 3 296 61
XM_017025756.1 XP_016881245.1 2 - 3 296 61
XM_017025757.1 XP_016881246.1 2 - 3 296 61
XM_017025758.1 XP_016881247.1 2 - 3 296 61
XM_017025759.1 XP_016881248.1 2 - 3 296 61
XM_017025760.1 XP_016881249.1 2 - 3 296 61
XM_017025761.1 XP_016881250.1 2 - 3 296 61
XM_017025762.1 XP_016881251.1 2 - 3 296 61
XM_017025763.1 XP_016881252.1 2 - 3 296 61
XM_017025764.1 XP_016881253.1 2 - 3 296 61
XM_017025765.1 XP_016881254.1 2 - 3 296 61
XM_017025766.1 XP_016881255.1 2 - 3 296 61
XM_017025767.1 XP_016881256.1 2 - 3 296 61
XM_017025768.1 XP_016881257.1 2 - 3 296 61
XM_017025769.1 XP_016881258.1 2 - 3 296 61
XM_017025770.1 XP_016881259.1 2 - 3 296 61
XM_017025771.1 XP_016881260.1 2 - 3 296 61
XM_017025772.1 XP_016881261.1 2 - 3 296 61
XM_017025773.1 XP_016881262.1 2 - 3 296 61
XM_017025774.1 XP_016881263.1 2 - 3 296 61
XM_017025775.1 XP_016881264.1 2 - 3 296 61
XM_017025776.1 XP_016881265.1 2 - 3 296 61
XM_017025777.1 XP_016881266.1 2 - 3 296 61
GenBank mRNA AF078830.1 - 1 - 2 112 61
AF078831.1 - 1 - 2 112 61
AF078832.1 - 1 - 2 112 61
AF078833.1 - 1 - 2 112 61
AJ564845.1 - 2 - 3 251 61
AK225889.1 - 2 - 3 459 61
AK225948.1 - 2 - 3 255 61
AK291519.1 - 2 - 3 550 61
AK295401.1 - 2 - 3 281 61
AK300759.1 - 2 - 3 330 61
AK302004.1 - 2 - 3 241 61
BC033242.1 - 2 - 3 275 61
BC047353.1 - 2 - 3 234 61
EF488685.1 - 1 - 2 112 61
Y10746.1 - 2 - 3 251 61

View Full Product Details