Assay Design



Gene Transcripts

Gene Symbol
MDM2
Entrez Gene ID
4193
Gene Name
MDM2 proto-oncogene
Gene Aliases
ACTFS, HDMX, hdm2
Chromosome Location
Chr.12: 68808149 - 68845544 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145337.2 NP_001138809.1 5 - 6 413 149
NM_002392.5 NP_002383.2 5 - 6 663 149
XM_005268872.4 XP_005268929.1 5 - 6 591 149
XM_006719399.3 XP_006719462.1 3 - 4 246 149
GenBank mRNA AF385323.1 - 4 - 5 339 149
AJ430612.1 - 4 - 5 339 149
AJ491698.1 - 4 - 5 339 149
AK290341.1 - 5 - 6 613 149
AM902642.1 - 5 - 6 432 149
AM902646.1 - 4 - 5 339 149
AM902650.1 - 4 - 5 339 149
AM902651.1 - 4 - 5 339 149
AM902654.1 - 4 - 5 339 149
AM902656.1 - 4 - 5 248 149
AM902658.1 - 5 - 6 426 149
AM902659.1 - 4 - 5 339 149
AM902666.1 - 5 - 6 384 194
AM902667.1 - 4 - 5 339 149
AM902668.1 - 4 - 5 339 149
AM902670.1 - 3 - 4 264 149
AM902672.1 - 4 - 5 339 149
BC152384.1 - 4 - 5 342 149
BC152390.1 - 4 - 5 342 149
BT007258.1 - 4 - 5 339 149
EU076747.1 - 5 - 6 413 149
EU076749.1 - 5 - 6 413 149
GQ848196.1 - 3 - 4 264 149
HM213901.1 - 4 - 5 339 149
JX015390.1 - 4 - 5 339 149
JX015391.1 - 4 - 5 339 149
JX015392.1 - 4 - 5 339 149
M92424.1 - 5 - 6 650 149
Z12020.1 - 5 - 6 650 149

View Full Product Details