Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004699.3 | NP_004690.1 | 1 - 2 | 216 | 67 |
GenBank mRNA | AD001530.1 | - | 1 - 2 | 181 | 67 |
AK293119.1 | - | 1 - 2 | 170 | 67 | |
AK313697.1 | - | 1 - 2 | 164 | 67 | |
AL365368.1 | - | 1 - 2 | 96 | 67 | |
AU143518.1 | - | 1 - 2 | 204 | 67 | |
BC000028.2 | - | 1 - 2 | 187 | 67 | |
CR992165.1 | - | 1 - 2 | 216 | 67 | |
D83260.1 | - | 1 - 2 | 86 | 67 |