Assay Design



Gene Transcripts

Gene Symbol
ZFYVE9
Entrez Gene ID
9372
Gene Name
zinc finger FYVE-type containing 9
Gene Aliases
MADHIP, NSP, PPP1R173, SARA, SMADIP
Chromosome Location
Chr.1: 52142001 - 52348664 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004799.3 NP_004790.2 5 - 6 2732 97
XM_011542437.2 XP_011540739.1 5 - 6 2603 97
XM_017002845.1 XP_016858334.1 5 - 6 2825 97
GenBank mRNA AB209125.1 - 2 - 3 2006 97
AF104304.1 - 4 - 5 2412 97
BC032680.1 - 3 - 4 2291 97

View Full Product Details