Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308028.1 | NP_001294957.1 | 9 - 10 | 1330 | 100 |
NM_001308031.1 | NP_001294960.1 | 2 - 3 | 280 | 100 | |
NM_005246.3 | NP_005237.2 | 11 - 12 | 1716 | 100 | |
XM_011543266.1 | XP_011541568.1 | 11 - 12 | 1569 | 100 | |
XM_011543267.1 | XP_011541569.1 | 11 - 12 | 3453 | 100 | |
XM_011543269.1 | XP_011541571.1 | 10 - 11 | 3307 | 100 | |
XM_011543270.2 | XP_011541572.1 | 10 - 11 | 1484 | 100 | |
XM_011543271.2 | XP_011541573.1 | 11 - 12 | 1569 | 100 | |
XM_011543276.2 | XP_011541578.1 | 2 - 3 | 487 | 100 | |
XM_017009229.1 | XP_016864718.1 | 12 - 13 | 1716 | 100 | |
XM_017009230.1 | XP_016864719.1 | 10 - 11 | 1484 | 100 | |
XM_017009231.1 | XP_016864720.1 | 10 - 11 | 3307 | 100 | |
XM_017009232.1 | XP_016864721.1 | 10 - 11 | 1357 | 100 | |
XM_017009233.1 | XP_016864722.1 | 9 - 10 | 1300 | 100 | |
XM_017009234.1 | XP_016864723.1 | 11 - 12 | 1569 | 100 | |
XM_017009235.1 | XP_016864724.1 | 11 - 12 | 1569 | 100 | |
GenBank mRNA | AK293376.1 | - | 8 - 9 | 1257 | 100 |
AK315234.1 | - | 9 - 10 | 1404 | 100 | |
BC017060.1 | - | 9 - 10 | 1398 | 100 | |
BU155168.1 | - | 2 - 3 | 289 | 100 | |
J03358.1 | - | 11 - 12 | 1716 | 100 |