Assay Design



Gene Transcripts

Gene Symbol
TPSD1
Entrez Gene ID
23430
Gene Name
tryptase delta 1
Gene Aliases
MCP7-LIKE, MCP7L1, MMCP-7L
Chromosome Location
Chr.16: 1256272 - 1258493 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012217.2 NP_036349.1 3 - 4 533 76
GenBank mRNA AF206664.1 - 2 - 3 370 76
BC069143.1 - 3 - 4 524 76

View Full Product Details