Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301089.1 | NP_001288018.1 | 7 - 8 | 977 | 68 |
NM_014758.2 | NP_055573.2 | 7 - 8 | 2991 | 68 | |
XM_005271546.4 | XP_005271603.1 | 7 - 8 | 2990 | 68 | |
XM_011542819.2 | XP_011541121.1 | 7 - 8 | 2990 | 68 | |
XM_011542820.2 | XP_011541122.1 | 7 - 8 | 2990 | 68 | |
XM_011542821.2 | XP_011541123.1 | 6 - 7 | 2871 | 68 | |
XM_011542822.2 | XP_011541124.1 | 7 - 8 | 2990 | 68 | |
XM_011542824.2 | XP_011541126.1 | 7 - 8 | 1895 | 68 | |
XM_011542825.2 | XP_011541127.1 | 7 - 8 | 1157 | 68 | |
XM_011542826.2 | XP_011541128.1 | 7 - 8 | 974 | 68 | |
XM_011542827.2 | XP_011541129.1 | 6 - 7 | 855 | 68 | |
XM_017017725.1 | XP_016873214.1 | 6 - 7 | 2846 | 68 | |
XM_017017726.1 | XP_016873215.1 | 7 - 8 | 1154 | 68 | |
GenBank mRNA | AF395843.1 | - | 7 - 8 | 2443 | 68 |
AK292457.1 | - | 7 - 8 | 3013 | 68 | |
AK294731.1 | - | 4 - 5 | 505 | 68 | |
AK304809.1 | - | 8 - 9 | 997 | 68 | |
AK308628.1 | - | 3 - 4 | 656 | 68 | |
AK316414.1 | - | 7 - 8 | 941 | 68 | |
AK316505.1 | - | 6 - 7 | 803 | 68 | |
D87443.1 | - | 7 - 8 | 2971 | 68 |