Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270764.1 | NP_001257693.1 | 7 - 8 | 2152 | 72 |
NM_015892.4 | NP_056976.2 | 7 - 8 | 2114 | 72 | |
XM_005269891.3 | XP_005269948.1 | 7 - 8 | 2230 | 72 | |
XM_006717891.3 | XP_006717954.1 | 7 - 8 | 2547 | 72 | |
XM_017016319.1 | XP_016871808.1 | 7 - 8 | 2066 | 72 | |
GenBank mRNA | AB011170.1 | - | 7 - 8 | 2082 | 72 |
AB062423.1 | - | 6 - 7 | 1501 | 72 | |
AF026477.1 | - | 6 - 7 | 1708 | 72 | |
BC027908.1 | - | 7 - 8 | 2057 | 72 | |
BC075813.1 | - | 6 - 7 | 1808 | 72 | |
CR749804.1 | - | 7 - 8 | 2235 | 72 |