Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014920.3 | NP_055735.1 | 1 - 2 | 206 | 95 |
XM_011514419.1 | XP_011512721.1 | 1 - 2 | 206 | 95 | |
GenBank mRNA | AB023153.1 | - | 1 - 2 | 93 | 95 |
AF152469.1 | - | 1 - 2 | 174 | 95 | |
BC152464.1 | - | 1 - 2 | 93 | 95 | |
DA456873.1 | - | 1 - 2 | 206 | 95 |