Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41257433-41257948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UCHL1
Entrez Gene ID
7345
Gene Name
ubiquitin C-terminal hydrolase L1
Gene Aliases
HEL-117, HEL-S-53, NDGOA, PARK5, PGP 9.5, PGP9.5, PGP95, Uch-L1
UniGene
Hs.518731
Gene Chromosome Location
Chr.4: 41256881-41268429 on Build GRCh38
Transcripts
NM_004181.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4159070 c.53C>A p.S18Y Chr.4: 41257616-41257616 on Build GRCh38
4738999 c.144G>A p.A48A Chr.4: 41257707-41257707 on Build GRCh38
4625295 c.127C>T p.P43S Chr.4: 41257690-41257690 on Build GRCh38
7254572 c.142G>T p.A48S Chr.4: 41257705-41257705 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs992824017 Chr.4: 41257808-41257808 on Build GRCh38
rs551675174 Chr.4: 41257845-41257845 on Build GRCh38
rs1011674126 Chr.4: 41257760-41257760 on Build GRCh38
rs565148227 Chr.4: 41257499-41257499 on Build GRCh38
rs1049115291 Chr.4: 41257576-41257576 on Build GRCh38
rs771225340 Chr.4: 41257730-41257731 on Build GRCh38
rs972692048 Chr.4: 41257552-41257552 on Build GRCh38
rs764704167 Chr.4: 41257769-41257769 on Build GRCh38
rs894583471 Chr.4: 41257594-41257594 on Build GRCh38
rs753345185 Chr.4: 41257566-41257566 on Build GRCh38
rs999158009 Chr.4: 41257648-41257648 on Build GRCh38
rs1045873735 Chr.4: 41257890-41257890 on Build GRCh38
rs1045884870 Chr.4: 41257626-41257626 on Build GRCh38
rs998869443 Chr.4: 41257864-41257864 on Build GRCh38
rs867177652 Chr.4: 41257685-41257685 on Build GRCh38
rs769782961 Chr.4: 41257726-41257726 on Build GRCh38
rs781765426 Chr.4: 41257713-41257713 on Build GRCh38
rs753720793 Chr.4: 41257632-41257632 on Build GRCh38
rs1029422646 Chr.4: 41257494-41257494 on Build GRCh38
rs892047129 Chr.4: 41257847-41257847 on Build GRCh38
rs186349613 Chr.4: 41257783-41257783 on Build GRCh38
rs772940357 Chr.4: 41257746-41257746 on Build GRCh38
rs375032754 Chr.4: 41257683-41257683 on Build GRCh38
rs767261015 Chr.4: 41257623-41257623 on Build GRCh38
rs556171924 Chr.4: 41257468-41257468 on Build GRCh38
rs747510594 Chr.4: 41257603-41257603 on Build GRCh38
rs1006625956 Chr.4: 41257754-41257754 on Build GRCh38
rs1044518948 Chr.4: 41257829-41257829 on Build GRCh38
rs1028034924 Chr.4: 41257765-41257765 on Build GRCh38
rs768922697 Chr.4: 41257740-41257740 on Build GRCh38
rs955752700 Chr.4: 41257460-41257460 on Build GRCh38
rs559788950 Chr.4: 41257544-41257544 on Build GRCh38
rs917282545 Chr.4: 41257809-41257809 on Build GRCh38
rs745957339 Chr.4: 41257617-41257617 on Build GRCh38
rs763182177 Chr.4: 41257619-41257619 on Build GRCh38
rs912437447 Chr.4: 41257788-41257788 on Build GRCh38
rs571904861 Chr.4: 41257870-41257870 on Build GRCh38
rs766097323 Chr.4: 41257762-41257762 on Build GRCh38
rs1049534502 Chr.4: 41257843-41257843 on Build GRCh38
rs372937139 Chr.4: 41257569-41257569 on Build GRCh38
rs368554502 Chr.4: 41257577-41257577 on Build GRCh38
rs560256408 Chr.4: 41257555-41257555 on Build GRCh38
rs972703444 Chr.4: 41257792-41257792 on Build GRCh38
rs763818311 Chr.4: 41257772-41257772 on Build GRCh38
rs752246660 Chr.4: 41257770-41257770 on Build GRCh38
rs778151038 Chr.4: 41257596-41257596 on Build GRCh38
rs758721522 Chr.4: 41257593-41257593 on Build GRCh38
rs919210760 Chr.4: 41257559-41257559 on Build GRCh38
rs773342302 Chr.4: 41257572-41257573 on Build GRCh38
rs953779930 Chr.4: 41257752-41257752 on Build GRCh38
rs889465372 Chr.4: 41257908-41257908 on Build GRCh38
rs930637351 Chr.4: 41257822-41257822 on Build GRCh38
rs769879042 Chr.4: 41257573-41257573 on Build GRCh38
rs915963124 Chr.4: 41257867-41257867 on Build GRCh38
rs766292262 Chr.4: 41257630-41257630 on Build GRCh38
rs868560463 Chr.4: 41257654-41257654 on Build GRCh38
rs936191539 Chr.4: 41257840-41257840 on Build GRCh38
rs960946980 Chr.4: 41257797-41257797 on Build GRCh38
rs747771091 Chr.4: 41257573-41257573 on Build GRCh38
rs749368841 Chr.4: 41257707-41257709 on Build GRCh38
rs531564323 Chr.4: 41257791-41257791 on Build GRCh38
rs867388308 Chr.4: 41257691-41257691 on Build GRCh38
rs372823328 Chr.4: 41257771-41257771 on Build GRCh38
rs532974616 Chr.4: 41257767-41257767 on Build GRCh38
rs370928430 Chr.4: 41257620-41257620 on Build GRCh38
rs551789430 Chr.4: 41257818-41257818 on Build GRCh38
rs758047620 Chr.4: 41257697-41257697 on Build GRCh38
rs548993481 Chr.4: 41257707-41257707 on Build GRCh38
rs769924890 Chr.4: 41257618-41257618 on Build GRCh38
rs986622298 Chr.4: 41257784-41257784 on Build GRCh38
rs985260501 Chr.4: 41257524-41257524 on Build GRCh38
rs541220563 Chr.4: 41257550-41257550 on Build GRCh38
rs760479916 Chr.4: 41257756-41257756 on Build GRCh38
rs1031726237 Chr.4: 41257459-41257459 on Build GRCh38
rs771560835 Chr.4: 41257605-41257605 on Build GRCh38
rs544988955 Chr.4: 41257490-41257490 on Build GRCh38
rs930709004 Chr.4: 41257547-41257547 on Build GRCh38
rs190746690 Chr.4: 41257884-41257884 on Build GRCh38
rs5030732 Chr.4: 41257616-41257616 on Build GRCh38
rs764766596 Chr.4: 41257662-41257662 on Build GRCh38
rs563836596 Chr.4: 41257831-41257831 on Build GRCh38
rs752315512 Chr.4: 41257678-41257678 on Build GRCh38
rs778064650 Chr.4: 41257701-41257701 on Build GRCh38
rs757635849 Chr.4: 41257711-41257711 on Build GRCh38
rs909743752 Chr.4: 41257538-41257538 on Build GRCh38
rs7658776 Chr.4: 41257920-41257920 on Build GRCh38
rs760676226 Chr.4: 41257628-41257628 on Build GRCh38
rs754490470 Chr.4: 41257657-41257657 on Build GRCh38
rs749516843 Chr.4: 41257731-41257731 on Build GRCh38
rs1016645998 Chr.4: 41257925-41257925 on Build GRCh38
rs891423143 Chr.4: 41257676-41257676 on Build GRCh38
rs112409507 Chr.4: 41257481-41257481 on Build GRCh38
rs754687056 Chr.4: 41257568-41257568 on Build GRCh38
rs953833361 Chr.4: 41257506-41257506 on Build GRCh38
rs929329652 Chr.4: 41257560-41257560 on Build GRCh38
rs547953322 Chr.4: 41257888-41257888 on Build GRCh38
rs746333982 Chr.4: 41257725-41257725 on Build GRCh38

View Full Product Details