Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41261627-41262133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UCHL1
Entrez Gene ID
7345
Gene Name
ubiquitin C-terminal hydrolase L1
Gene Aliases
HEL-117, HEL-S-53, NDGOA, PARK5, PGP 9.5, PGP9.5, PGP95, Uch-L1
UniGene
Hs.518731
Gene Chromosome Location
Chr.4: 41256881-41268429 on Build GRCh38
Transcripts
NM_004181.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4739000 c.352C>A p.L118I Chr.4: 41261741-41261741 on Build GRCh38
297787 c.435C>T p.A145A Chr.4: 41261899-41261899 on Build GRCh38
6469557 c.418C>G p.Q140E Chr.4: 41261882-41261882 on Build GRCh38
3696648 c.457C>T p.R153W Chr.4: 41261921-41261921 on Build GRCh38
1055378 c.458G>A p.R153Q Chr.4: 41261922-41261922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200793891 Chr.4: 41261960-41261960 on Build GRCh38
rs763168187 Chr.4: 41261812-41261813 on Build GRCh38
rs368441837 Chr.4: 41261711-41261711 on Build GRCh38
rs924143390 Chr.4: 41261868-41261868 on Build GRCh38
rs199514166 Chr.4: 41261926-41261926 on Build GRCh38
rs36070748 Chr.4: 41262087-41262087 on Build GRCh38
rs548383480 Chr.4: 41262014-41262014 on Build GRCh38
rs181585116 Chr.4: 41261937-41261937 on Build GRCh38
rs573101519 Chr.4: 41262032-41262032 on Build GRCh38
rs139583787 Chr.4: 41261921-41261921 on Build GRCh38
rs867149026 Chr.4: 41261735-41261735 on Build GRCh38
rs766323147 Chr.4: 41261819-41261819 on Build GRCh38
rs777973619 Chr.4: 41261786-41261786 on Build GRCh38
rs761194133 Chr.4: 41261734-41261735 on Build GRCh38
rs761622006 Chr.4: 41261692-41261692 on Build GRCh38
rs759897414 Chr.4: 41261946-41261946 on Build GRCh38
rs553207196 Chr.4: 41261998-41261998 on Build GRCh38
rs747678137 Chr.4: 41261823-41261823 on Build GRCh38
rs200775509 Chr.4: 41261792-41261792 on Build GRCh38
rs886238721 Chr.4: 41261905-41261905 on Build GRCh38
rs760197398 Chr.4: 41261851-41261851 on Build GRCh38
rs754392787 Chr.4: 41261753-41261753 on Build GRCh38
rs774059700 Chr.4: 41261820-41261820 on Build GRCh38
rs771646598 Chr.4: 41261832-41261832 on Build GRCh38
rs397719664 Chr.4: 41261711-41261712 on Build GRCh38
rs992319581 Chr.4: 41261663-41261663 on Build GRCh38
rs779687111 Chr.4: 41261808-41261808 on Build GRCh38
rs764784241 Chr.4: 41261900-41261900 on Build GRCh38
rs758414333 Chr.4: 41261777-41261777 on Build GRCh38
rs753050379 Chr.4: 41261863-41261864 on Build GRCh38
rs141828940 Chr.4: 41261745-41261745 on Build GRCh38
rs763427877 Chr.4: 41261965-41261965 on Build GRCh38
rs542014346 Chr.4: 41262078-41262078 on Build GRCh38
rs771734476 Chr.4: 41261922-41261922 on Build GRCh38
rs759399692 Chr.4: 41261678-41261679 on Build GRCh38
rs3214812 Chr.4: 41261701-41261702 on Build GRCh38
rs376624356 Chr.4: 41261772-41261772 on Build GRCh38
rs752770640 Chr.4: 41261774-41261774 on Build GRCh38
rs375810834 Chr.4: 41262016-41262016 on Build GRCh38
rs370581648 Chr.4: 41261814-41261814 on Build GRCh38
rs747159120 Chr.4: 41261790-41261790 on Build GRCh38
rs753134305 Chr.4: 41261878-41261878 on Build GRCh38
rs779001647 Chr.4: 41261919-41261919 on Build GRCh38
rs139618482 Chr.4: 41261796-41261796 on Build GRCh38
rs918013729 Chr.4: 41262066-41262066 on Build GRCh38
rs781480182 Chr.4: 41261837-41261838 on Build GRCh38
rs766816754 Chr.4: 41261861-41261861 on Build GRCh38
rs764985626 Chr.4: 41261813-41261814 on Build GRCh38
rs757499033 Chr.4: 41262022-41262022 on Build GRCh38
rs767268217 Chr.4: 41261699-41261699 on Build GRCh38
rs755399037 Chr.4: 41261838-41261839 on Build GRCh38
rs772943928 Chr.4: 41261711-41261711 on Build GRCh38
rs894419016 Chr.4: 41262090-41262090 on Build GRCh38
rs1048744513 Chr.4: 41261995-41261995 on Build GRCh38
rs147393026 Chr.4: 41261908-41261908 on Build GRCh38
rs753465403 Chr.4: 41261704-41261705 on Build GRCh38
rs751971352 Chr.4: 41261952-41261952 on Build GRCh38
rs1036219653 Chr.4: 41261684-41261684 on Build GRCh38
rs773966040 Chr.4: 41261665-41261665 on Build GRCh38
rs764111400 Chr.4: 41261887-41261887 on Build GRCh38
rs372479324 Chr.4: 41261736-41261736 on Build GRCh38
rs557049900 Chr.4: 41261917-41261917 on Build GRCh38
rs777112600 Chr.4: 41261943-41261943 on Build GRCh38
rs772952946 Chr.4: 41261842-41261842 on Build GRCh38
rs749021086 Chr.4: 41261914-41261914 on Build GRCh38
rs776832748 Chr.4: 41261865-41261865 on Build GRCh38
rs756685050 Chr.4: 41261869-41261870 on Build GRCh38
rs1051597786 Chr.4: 41261901-41261901 on Build GRCh38
rs374298650 Chr.4: 41261871-41261871 on Build GRCh38
rs555081806 Chr.4: 41262081-41262081 on Build GRCh38
rs749992093 Chr.4: 41261835-41261836 on Build GRCh38
rs758504465 Chr.4: 41261879-41261879 on Build GRCh38
rs141609433 Chr.4: 41262049-41262049 on Build GRCh38
rs907106072 Chr.4: 41261688-41261688 on Build GRCh38
rs764271024 Chr.4: 41261975-41261975 on Build GRCh38
rs565798637 Chr.4: 41261702-41261703 on Build GRCh38
rs765745344 Chr.4: 41261872-41261872 on Build GRCh38
rs548705677 Chr.4: 41261810-41261810 on Build GRCh38
rs371322251 Chr.4: 41261942-41261942 on Build GRCh38
rs11556274 Chr.4: 41261742-41261742 on Build GRCh38
rs142811772 Chr.4: 41261899-41261899 on Build GRCh38
rs1012642269 Chr.4: 41262099-41262099 on Build GRCh38
rs150601238 Chr.4: 41261759-41261759 on Build GRCh38
rs796640089 Chr.4: 41261702-41261703 on Build GRCh38
rs753316777 Chr.4: 41261689-41261689 on Build GRCh38
rs907037378 Chr.4: 41262050-41262050 on Build GRCh38
rs900235906 Chr.4: 41261889-41261889 on Build GRCh38

View Full Product Details