Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41268054-41268575 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UCHL1
Entrez Gene ID
7345
Gene Name
ubiquitin C-terminal hydrolase L1
Gene Aliases
HEL-117, HEL-S-53, NDGOA, PARK5, PGP 9.5, PGP9.5, PGP95, Uch-L1
UniGene
Hs.518731
Gene Chromosome Location
Chr.4: 41256881-41268429 on Build GRCh38
Transcripts
NM_004181.4

SNP Detail

SNP ID Chromosome Location
rs773158014 Chr.4: 41268094-41268094 on Build GRCh38
rs1004149787 Chr.4: 41268331-41268331 on Build GRCh38
rs760542056 Chr.4: 41268131-41268132 on Build GRCh38
rs921700740 Chr.4: 41268491-41268491 on Build GRCh38
rs924938811 Chr.4: 41268445-41268445 on Build GRCh38
rs886389553 Chr.4: 41268329-41268329 on Build GRCh38
rs11556270 Chr.4: 41268313-41268313 on Build GRCh38
rs140683078 Chr.4: 41268318-41268318 on Build GRCh38
rs1025217461 Chr.4: 41268406-41268406 on Build GRCh38
rs768753947 Chr.4: 41268143-41268143 on Build GRCh38
rs370431599 Chr.4: 41268086-41268086 on Build GRCh38
rs977780391 Chr.4: 41268251-41268251 on Build GRCh38
rs959534314 Chr.4: 41268157-41268157 on Build GRCh38
rs933142689 Chr.4: 41268511-41268511 on Build GRCh38
rs1051572084 Chr.4: 41268531-41268531 on Build GRCh38
rs569065797 Chr.4: 41268467-41268468 on Build GRCh38
rs936469594 Chr.4: 41268296-41268296 on Build GRCh38
rs755024590 Chr.4: 41268403-41268403 on Build GRCh38
rs553172248 Chr.4: 41268151-41268151 on Build GRCh38
rs116336893 Chr.4: 41268349-41268349 on Build GRCh38
rs966991717 Chr.4: 41268421-41268421 on Build GRCh38
rs759170558 Chr.4: 41268136-41268136 on Build GRCh38
rs758867690 Chr.4: 41268140-41268140 on Build GRCh38
rs115323706 Chr.4: 41268307-41268307 on Build GRCh38
rs752225914 Chr.4: 41268139-41268139 on Build GRCh38
rs776684041 Chr.4: 41268145-41268146 on Build GRCh38
rs535206351 Chr.4: 41268300-41268300 on Build GRCh38
rs207464559 Chr.4: 41268435-41268435 on Build GRCh38
rs77194345 Chr.4: 41268093-41268093 on Build GRCh38
rs560407112 Chr.4: 41268359-41268359 on Build GRCh38
rs555393576 Chr.4: 41268302-41268302 on Build GRCh38
rs765917631 Chr.4: 41268119-41268119 on Build GRCh38
rs760586461 Chr.4: 41268117-41268117 on Build GRCh38
rs778936444 Chr.4: 41268437-41268437 on Build GRCh38
rs918190793 Chr.4: 41268199-41268199 on Build GRCh38
rs564857603 Chr.4: 41268323-41268323 on Build GRCh38
rs925022579 Chr.4: 41268284-41268284 on Build GRCh38
rs895198680 Chr.4: 41268198-41268198 on Build GRCh38
rs78683791 Chr.4: 41268382-41268382 on Build GRCh38
rs898564250 Chr.4: 41268362-41268362 on Build GRCh38
rs540507296 Chr.4: 41268357-41268357 on Build GRCh38
rs778259499 Chr.4: 41268142-41268142 on Build GRCh38
rs974642727 Chr.4: 41268091-41268091 on Build GRCh38
rs766043762 Chr.4: 41268460-41268460 on Build GRCh38
rs16852986 Chr.4: 41268454-41268454 on Build GRCh38
rs978154363 Chr.4: 41268424-41268424 on Build GRCh38
rs116700032 Chr.4: 41268367-41268367 on Build GRCh38
rs183586019 Chr.4: 41268204-41268204 on Build GRCh38
rs530842282 Chr.4: 41268522-41268522 on Build GRCh38
rs886059408 Chr.4: 41268198-41268198 on Build GRCh38
rs944994122 Chr.4: 41268250-41268250 on Build GRCh38
rs386673866 Chr.4: 41268250-41268251 on Build GRCh38
rs369814359 Chr.4: 41268090-41268090 on Build GRCh38
rs369393584 Chr.4: 41268427-41268427 on Build GRCh38
rs1034182930 Chr.4: 41268377-41268377 on Build GRCh38
rs753389158 Chr.4: 41268133-41268133 on Build GRCh38
rs974497092 Chr.4: 41268485-41268485 on Build GRCh38
rs775513694 Chr.4: 41268075-41268075 on Build GRCh38
rs992371452 Chr.4: 41268161-41268161 on Build GRCh38

View Full Product Details