Assay Design



Gene Transcripts

Gene Symbol
TAS2R4
Entrez Gene ID
50832
Gene Name
taste 2 receptor member 4
Gene Aliases
T2R4
Chromosome Location
Chr.7: 141778489 - 141779388 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016944.1 NP_058640.1 1 - 1 451 66
GenBank mRNA BC130439.1 - 1 - 1 498 66
BC136508.1 - 1 - 1 498 66

View Full Product Details