Assay Design



Gene Transcripts

Gene Symbol
MFAP2
Entrez Gene ID
4237
Gene Name
microfibrillar associated protein 2
Gene Aliases
MAGP, MAGP-1, MAGP1
Chromosome Location
Chr.1: 16974502 - 16981649 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002403.3 NP_002394.1 2 - 3 174 115
NM_017459.2 NP_059453.1 2 - 3 155 115
XM_011541476.2 XP_011539778.1 2 - 3 781 115
GenBank mRNA AK301655.1 - 2 - 3 147 115
AU280150.1 - 2 - 3 175 115
BC015039.1 - 2 - 3 174 115
CR407678.1 - 1 - 2 41 115
DN996554.1 - 2 - 3 124 115
U19718.1 - 1 - 2 82 115

View Full Product Details