Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190266.1 | NP_001177195.1 | 9 - 10 | 1209 | 69 |
NM_001190267.1 | NP_001177196.1 | 9 - 10 | 1207 | 69 | |
NM_017974.3 | NP_060444.3 | 8 - 9 | 1154 | 69 | |
NM_030803.6 | NP_110430.5 | 9 - 10 | 1211 | 69 | |
NM_198890.2 | NP_942593.2 | 5 - 6 | 722 | 69 | |
XM_005246082.2 | XP_005246139.1 | 10 - 11 | 1199 | 69 | |
XM_005246084.2 | XP_005246141.1 | 7 - 8 | 738 | 69 | |
XM_005246086.2 | XP_005246143.1 | 6 - 7 | 687 | 69 | |
XM_006712608.2 | XP_006712671.1 | 9 - 10 | 1010 | 69 | |
GenBank mRNA | AK000897.1 | - | 8 - 9 | 822 | 69 |
AK001690.1 | - | 8 - 9 | 1000 | 69 | |
AK024453.1 | - | 8 - 9 | 1749 | 69 | |
AK074434.1 | - | 3 - 4 | 181 | 69 | |
AK123876.1 | - | 6 - 7 | 678 | 69 | |
AK308400.1 | - | 8 - 9 | 1052 | 69 | |
AY358182.1 | - | 9 - 10 | 1100 | 69 | |
AY398617.1 | - | 9 - 10 | 1100 | 69 | |
BC071846.1 | - | 5 - 6 | 560 | 69 | |
BC117337.1 | - | 9 - 10 | 873 | 69 | |
BC143900.1 | - | 8 - 9 | 816 | 69 | |
EF079889.1 | - | 8 - 9 | 897 | 69 | |
EF079890.1 | - | 5 - 6 | 465 | 69 | |
EF560734.1 | - | 8 - 9 | 1024 | 69 | |
JQ924062.1 | - | 8 - 9 | 897 | 69 |