Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018460.3 | NP_060930.3 | 7 - 8 | 725 | 73 |
XM_011511479.2 | XP_011509781.1 | 8 - 9 | 757 | 73 | |
XM_011511481.2 | XP_011509783.1 | 7 - 8 | 660 | 73 | |
XM_011511482.2 | XP_011509784.1 | 7 - 8 | 660 | 73 | |
XM_011511483.1 | XP_011509785.1 | 3 - 4 | 355 | 73 | |
XM_011511484.1 | XP_011509786.1 | 7 - 8 | 660 | 73 | |
XM_017004499.1 | XP_016859988.1 | 7 - 8 | 686 | 73 | |
XM_017004500.1 | XP_016859989.1 | 6 - 7 | 552 | 73 | |
XM_017004501.1 | XP_016859990.1 | 7 - 8 | 660 | 73 | |
GenBank mRNA | AF212222.1 | - | 7 - 8 | 663 | 73 |
AF217522.1 | - | 7 - 8 | 662 | 73 | |
AK226014.1 | - | 7 - 8 | 660 | 73 | |
AK303490.1 | - | 7 - 8 | 643 | 73 | |
AK308219.1 | - | 7 - 8 | 683 | 73 | |
AY219338.1 | - | 7 - 8 | 663 | 73 | |
BC016701.2 | - | 7 - 8 | 631 | 73 | |
BC038976.1 | - | 7 - 8 | 630 | 73 |