Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 89726160-89726659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNCA
Entrez Gene ID
6622
Gene Name
synuclein alpha
Gene Aliases
NACP, PARK1, PARK4, PD1
UniGene
Hs.21374
Gene Chromosome Location
Chr.4: 89724099-89838324 on Build GRCh38
Transcripts
NM_000345.3 NM_001146054.1 NM_001146055.1 NM_007308.2 XM_011532208.2 XM_017008562.1 XM_017008563.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5461644 c.400C>A p.Q134K Chr.4: 89726651-89726651 on Build GRCh38
2957802 c.405C>A p.D135E Chr.4: 89726646-89726646 on Build GRCh38
6469769 c.408C>T p.Y136Y Chr.4: 89726643-89726643 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754479691 Chr.4: 89726590-89726590 on Build GRCh38
rs778588756 Chr.4: 89726605-89726605 on Build GRCh38
rs982618263 Chr.4: 89726267-89726267 on Build GRCh38
rs1030477649 Chr.4: 89726523-89726523 on Build GRCh38
rs955311825 Chr.4: 89726532-89726532 on Build GRCh38
rs567582133 Chr.4: 89726205-89726205 on Build GRCh38
rs1043452934 Chr.4: 89726520-89726520 on Build GRCh38
rs962391035 Chr.4: 89726565-89726565 on Build GRCh38
rs549732620 Chr.4: 89726315-89726315 on Build GRCh38
rs748631342 Chr.4: 89726516-89726516 on Build GRCh38
rs560702695 Chr.4: 89726510-89726510 on Build GRCh38
rs373801428 Chr.4: 89726547-89726547 on Build GRCh38
rs35733299 Chr.4: 89726252-89726252 on Build GRCh38
rs192194319 Chr.4: 89726456-89726456 on Build GRCh38
rs918500594 Chr.4: 89726377-89726377 on Build GRCh38
rs973701461 Chr.4: 89726586-89726586 on Build GRCh38
rs1006605158 Chr.4: 89726544-89726544 on Build GRCh38
rs985546471 Chr.4: 89726223-89726223 on Build GRCh38
rs973920574 Chr.4: 89726359-89726359 on Build GRCh38
rs948134712 Chr.4: 89726453-89726453 on Build GRCh38
rs578250733 Chr.4: 89726466-89726466 on Build GRCh38
rs755913681 Chr.4: 89726578-89726578 on Build GRCh38
rs755608841 Chr.4: 89726585-89726585 on Build GRCh38
rs886059727 Chr.4: 89726351-89726351 on Build GRCh38
rs1052723475 Chr.4: 89726495-89726495 on Build GRCh38
rs955134665 Chr.4: 89726222-89726222 on Build GRCh38
rs962129321 Chr.4: 89726353-89726353 on Build GRCh38
rs79726680 Chr.4: 89726455-89726455 on Build GRCh38
rs755315517 Chr.4: 89726305-89726305 on Build GRCh38
rs370319708 Chr.4: 89726545-89726545 on Build GRCh38
rs144511886 Chr.4: 89726512-89726512 on Build GRCh38
rs553599518 Chr.4: 89726379-89726379 on Build GRCh38
rs34825021 Chr.4: 89726434-89726434 on Build GRCh38
rs941282317 Chr.4: 89726256-89726256 on Build GRCh38
rs779277002 Chr.4: 89726584-89726584 on Build GRCh38
rs571218440 Chr.4: 89726346-89726346 on Build GRCh38
rs193289281 Chr.4: 89726355-89726355 on Build GRCh38
rs976242802 Chr.4: 89726203-89726203 on Build GRCh38
rs35716318 Chr.4: 89726425-89726425 on Build GRCh38
rs896440399 Chr.4: 89726563-89726563 on Build GRCh38
rs1034908610 Chr.4: 89726238-89726238 on Build GRCh38
rs565855781 Chr.4: 89726420-89726420 on Build GRCh38
rs145304567 Chr.4: 89726551-89726551 on Build GRCh38
rs986792244 Chr.4: 89726488-89726488 on Build GRCh38
rs1031179031 Chr.4: 89726190-89726190 on Build GRCh38
rs886059728 Chr.4: 89726417-89726417 on Build GRCh38
rs781595227 Chr.4: 89726632-89726632 on Build GRCh38
rs187958987 Chr.4: 89726454-89726454 on Build GRCh38
rs1002542905 Chr.4: 89726235-89726238 on Build GRCh38
rs757748949 Chr.4: 89726631-89726631 on Build GRCh38
rs778443752 Chr.4: 89726572-89726572 on Build GRCh38
rs185476145 Chr.4: 89726426-89726426 on Build GRCh38
rs747474608 Chr.4: 89726609-89726609 on Build GRCh38
rs10024743 Chr.4: 89726489-89726489 on Build GRCh38
rs772716317 Chr.4: 89726517-89726517 on Build GRCh38
rs753544938 Chr.4: 89726588-89726588 on Build GRCh38
rs184023281 Chr.4: 89726511-89726511 on Build GRCh38
rs956897832 Chr.4: 89726196-89726196 on Build GRCh38

View Full Product Details