Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019106.5 | NP_061979.3 | 3 - 4 | 470 | 77 |
NM_145733.2 | NP_663786.2 | 3 - 4 | 470 | 77 | |
XM_017028862.1 | XP_016884351.1 | 4 - 5 | 2068 | 77 | |
GenBank mRNA | AF285107.1 | - | 3 - 4 | 347 | 77 |
AF285109.1 | - | 3 - 4 | 347 | 77 | |
AK290037.1 | - | 3 - 4 | 455 | 77 | |
AK299919.1 | - | 2 - 3 | 215 | 77 | |
AL833942.1 | - | 3 - 4 | 405 | 77 | |
BC111779.1 | - | 3 - 4 | 363 | 77 | |
CR456572.1 | - | 3 - 4 | 359 | 77 |