Assay Design
Gene Transcripts
- Gene Symbol
- ANKS1B
- Entrez Gene ID
-
56899
- Gene Name
- ankyrin repeat and sterile alpha motif domain containing 1B
- Gene Aliases
-
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
- Chromosome Location
-
Chr.12: 98729904 - 99984727 on Build GRCh38
View Full Product Details