Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018070.2 | NP_001018080.1 | 3 - 4 | 336 | 92 |
NM_020441.2 | NP_065174.1 | 2 - 3 | 314 | 92 | |
GenBank mRNA | AK000860.1 | - | 2 - 3 | 297 | 92 |
AK295782.1 | - | 2 - 3 | 274 | 92 | |
AK309267.1 | - | 2 - 3 | 289 | 92 | |
AK315399.1 | - | 2 - 3 | 269 | 92 | |
BC006449.1 | - | 2 - 3 | 264 | 92 | |
BI767141.1 | - | 3 - 4 | 312 | 92 | |
DA061767.1 | - | 3 - 4 | 336 | 92 |