Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020992.3 | NP_066272.1 | 4 - 5 | 769 | 135 |
XM_011540330.1 | XP_011538632.1 | 3 - 4 | 475 | 135 | |
GenBank mRNA | AJ310549.1 | - | 4 - 5 | 536 | 135 |
AK314792.1 | - | 4 - 5 | 135 | ||
BC000915.2 | - | 4 - 5 | 645 | 135 | |
BC018755.1 | - | 4 - 5 | 560 | 135 | |
FJ224331.1 | - | 4 - 5 | 560 | 135 | |
U90878.1 | - | 4 - 5 | 678 | 135 |