Assay Design



Gene Transcripts

Gene Symbol
CXorf56
Entrez Gene ID
63932
Gene Name
chromosome X open reading frame 56
Gene Aliases
-
Chromosome Location
Chr.X: 119538149 - 119565434 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170569.1 NP_001164040.1 2 - 3 404 93
NM_022101.3 NP_071384.1 2 - 3 322 93
GenBank mRNA AK096297.1 - 2 - 3 368 93
BC023506.2 - 2 - 3 283 93
BQ223940.1 - 2 - 3 287 93
DA590933.1 - 2 - 3 406 93
DA921725.1 - 2 - 3 297 93
DB033119.1 - 2 - 3 337 93

View Full Product Details