Assay Design



Gene Transcripts

Gene Symbol
CDH23
Entrez Gene ID
64072
Gene Name
cadherin-related 23
Gene Aliases
CDHR23, USH1D
Chromosome Location
Chr.10: 71396934 - 71815947 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171930.1 NP_001165401.1 2 - 3 458 96
NM_001171931.1 NP_001165402.1 2 - 3 458 96
NM_001171932.1 NP_001165403.1 2 - 3 458 96
NM_022124.5 NP_071407.4 2 - 3 458 96
NM_052836.3 NP_443068.1 2 - 3 458 96
XM_006717940.3 XP_006718003.1 2 - 3 452 96
XM_006717942.3 XP_006718005.1 2 - 3 452 96
XM_011540039.2 XP_011538341.1 2 - 3 452 96
XM_011540042.2 XP_011538344.1 2 - 3 452 96
XM_011540043.2 XP_011538345.1 2 - 3 452 96
XM_011540044.2 XP_011538346.1 2 - 3 93 96
XM_011540045.2 XP_011538347.1 2 - 3 452 96
XM_011540048.2 XP_011538350.1 2 - 3 452 96
XM_011540049.2 XP_011538351.1 2 - 3 452 96
XM_011540051.2 XP_011538353.1 2 - 3 452 96
XM_017016499.1 XP_016871988.1 2 - 3 458 96
XM_017016500.1 XP_016871989.1 2 - 3 452 96
GenBank mRNA AF312024.1 - 1 - 2 68 96
AY010111.2 - 1 - 2 68 96
AY358617.1 - 2 - 3 108 96
BC011570.1 - 2 - 3 463 96
BC032581.1 - 2 - 3 464 96
BC065284.1 - 2 - 3 435 96
BC136976.1 - 2 - 3 328 96
BC136977.1 - 2 - 3 327 96
BC139903.1 - 2 - 3 450 96

View Full Product Details