Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003319.4 | NP_003310.4 | 46 - 47 | 13513 | 72 |
NM_133432.3 | NP_597676.3 | 47 - 48 | 13888 | 72 | |
NM_133437.4 | NP_597681.4 | 47 - 48 | 14089 | 72 | |
XM_017004823.1 | XP_016860312.1 | 47 - 48 | 13654 | 72 | |
GenBank mRNA | X90568.1 | - | 46 - 47 | 13420 | 72 |