Assay Design
Gene Transcripts
- Gene Symbol
- ABCF2
- Entrez Gene ID
-
10061
- Gene Name
- ATP binding cassette subfamily F member 2
- Gene Aliases
-
ABC28, EST133090, HUSSY-18, HUSSY18
- Chromosome Location
-
Chr.7: 151207837 - 151227374 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_005692.4 |
NP_005683.2 |
15 - 16 |
2121 |
99 |
GenBank mRNA |
AL050291.1 |
- |
14 - 15 |
1913 |
99 |
View Full Product Details