Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006872.4 | NP_006863.2 | 1 - 2 | 119 | 73 |
GenBank mRNA | AF106857.1 | - | 1 - 2 | 39 | 73 |
AK223350.1 | - | 1 - 2 | 43 | 73 | |
BC025991.1 | - | 1 - 2 | 68 | 73 | |
BG717289.1 | - | 1 - 2 | 95 | 73 | |
HY049561.1 | - | 1 - 2 | 122 | 73 |