Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271723.1 | NP_001258652.1 | 5 - 6 | 760 | 99 |
NM_030793.4 | NP_110420.3 | 5 - 6 | 760 | 99 | |
XM_005268513.1 | XP_005268570.1 | 5 - 6 | 749 | 99 | |
XM_006714797.1 | XP_006714860.1 | 5 - 6 | 749 | 99 | |
XM_011537684.2 | XP_011535986.1 | 4 - 5 | 655 | 99 | |
XM_017009899.1 | XP_016865388.1 | 5 - 6 | 738 | 99 | |
XM_017009900.1 | XP_016865389.1 | 4 - 5 | 655 | 99 | |
XM_017009901.1 | XP_016865390.1 | 4 - 5 | 655 | 99 | |
XM_017009902.1 | XP_016865391.1 | 4 - 5 | 655 | 99 | |
GenBank mRNA | AK024024.1 | - | 5 - 6 | 680 | 99 |
AK308414.1 | - | 5 - 6 | 708 | 99 | |
BC033454.1 | - | 5 - 6 | 693 | 99 | |
BC050424.1 | - | 4 - 5 | 665 | 99 |