Assay Design
Gene Transcripts
- Gene Symbol
- TRMT44
- Entrez Gene ID
-
152992
- Gene Name
- tRNA methyltransferase 44 homolog (S. cerevisiae)
- Gene Aliases
-
C4orf23, METTL19, TRM44
- Chromosome Location
-
Chr.4: 8440774 - 8512531 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK022953.1 |
- |
1 - 1 |
970 |
128 |
AK092387.1 |
- |
2 - 2 |
2908 |
128 |
AK096417.1 |
- |
1 - 1 |
2287 |
128 |
View Full Product Details