Assay Design



Gene Transcripts

Gene Symbol
SRSF8
Entrez Gene ID
10929
Gene Name
serine and arginine rich splicing factor 8
Gene Aliases
DSM-1, SFRS2B, SRP46
Chromosome Location
Chr.11: 95066877 - 95071227 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032102.3 NP_115285.1 1 - 1 1446 132
XM_017017142.1 XP_016872631.1 1 - 1 1408 132
GenBank mRNA AF031166.1 - 1 - 1 1066 132
AK309776.1 - 3 - 3 758 132
BC057783.1 - 2 - 2 1136 132
CB215027.1 - 1 - 1 407 132

View Full Product Details