Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002588.3 | NP_002579.2 | 1 - 1 | 2095 | 112 |
NM_032402.1 | NP_115778.1 | 1 - 1 | 2095 | 112 | |
GenBank mRNA | AF152337.1 | - | 1 - 1 | 1980 | 112 |
AF152524.1 | - | 1 - 1 | 1980 | 112 | |
BC019299.1 | - | 1 - 1 | 2094 | 112 | |
BC026218.1 | - | 1 - 1 | 2084 | 112 | |
L11373.1 | - | 1 - 1 | 2091 | 112 |