Assay Design
Gene Transcripts
- Gene Symbol
- TTTY6B
- Entrez Gene ID
-
441543
- Gene Name
- testis-specific transcript, Y-linked 6B (non-protein coding)
- Gene Aliases
-
LINC00128, NCRNA00128, TTTY6, TTY6
- Chromosome Location
-
Chr.Y: 22144987 - 22146834 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002175.3 |
|
2 - 3 |
|
118 |
GenBank mRNA |
AA884105.1 |
- |
1 - 2 |
245 |
118 |
- Gene Symbol
- TTTY6
- Entrez Gene ID
-
84672
- Gene Name
- testis-specific transcript, Y-linked 6 (non-protein coding)
- Gene Aliases
-
LINC00127, NCRNA00127, TTTY6A, TTY6
- Chromosome Location
-
Chr.Y: 22439590 - 22441459 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_001527.2 |
|
2 - 3 |
|
118 |
View Full Product Details