Assay Design



Gene Transcripts

Gene Symbol
MYO18B
Entrez Gene ID
84700
Gene Name
myosin XVIIIB
Gene Aliases
KFS4
Chromosome Location
Chr.22: 25742133 - 26063847 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318245.1 NP_001305174.1 12 - 13 2780 78
NM_032608.6 NP_115997.5 12 - 13 2780 78
XM_011530458.2 XP_011528760.1 13 - 14 2916 78
XM_011530459.2 XP_011528761.1 13 - 14 2887 78
XM_011530460.2 XP_011528762.1 12 - 13 2760 78
XM_011530461.2 XP_011528763.1 13 - 14 2916 78
XM_011530464.2 XP_011528766.1 13 - 14 2916 78
XM_011530465.2 XP_011528767.1 12 - 13 2673 78
XM_011530466.2 XP_011528768.1 13 - 14 2917 78
XM_017029012.1 XP_016884501.1 13 - 14 2916 78
XM_017029013.1 XP_016884502.1 13 - 14 2964 78
XM_017029014.1 XP_016884503.1 13 - 14 2916 78
XM_017029015.1 XP_016884504.1 12 - 13 2790 78
GenBank mRNA AB042648.2 - 8 - 9 1092 78
AB075376.1 - 12 - 13 2780 78
AJ310931.2 - 12 - 13 2771 78
AY077700.1 - 12 - 13 2690 78

View Full Product Details