Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032609.2 | NP_115998.2 | 3 - 4 | 327 | 59 |
XM_005260579.4 | XP_005260636.1 | 2 - 3 | 329 | 59 | |
GenBank mRNA | AF257180.1 | - | 3 - 4 | 327 | 59 |
BC057779.1 | - | 3 - 4 | 303 | 59 | |
HE647859.1 | - | 2 - 3 | 252 | 59 |