Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321831.1 | NP_001308760.1 | 11 - 12 | 1313 | 80 |
NM_139239.2 | NP_640332.1 | 11 - 12 | 1476 | 80 | |
XM_005259340.3 | XP_005259397.1 | 11 - 12 | 1692 | 80 | |
XM_011527419.2 | XP_011525721.1 | 11 - 12 | 1364 | 80 | |
XM_011527420.2 | XP_011525722.1 | 11 - 12 | 1375 | 80 | |
XM_011527421.2 | XP_011525723.1 | 10 - 11 | 1208 | 80 | |
XM_011527424.2 | XP_011525726.1 | 11 - 12 | 1320 | 80 | |
XM_017027390.1 | XP_016882879.1 | 11 - 12 | 1674 | 80 | |
XM_017027391.1 | XP_016882880.1 | 11 - 12 | 1627 | 80 | |
XM_017027392.1 | XP_016882881.1 | 11 - 12 | 1272 | 80 | |
XM_017027393.1 | XP_016882882.1 | 11 - 12 | 1271 | 80 | |
XM_017027394.1 | XP_016882883.1 | 9 - 10 | 1519 | 80 | |
XM_017027395.1 | XP_016882884.1 | 9 - 10 | 1501 | 80 | |
XM_017027397.1 | XP_016882886.1 | 9 - 10 | 1458 | 80 | |
GenBank mRNA | AF385433.1 | - | 11 - 12 | 1366 | 80 |
AF385434.1 | - | 11 - 12 | 1464 | 80 | |
BC006273.2 | - | 5 - 6 | 613 | 80 | |
BC119646.1 | - | 8 - 9 | 988 | 80 | |
BC119647.1 | - | 8 - 9 | 988 | 80 |