Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032836.2 | NP_116225.2 | 2 - 3 | 386 | 67 |
XM_005259352.4 | XP_005259409.1 | 2 - 3 | 1411 | 67 | |
GenBank mRNA | AK027674.1 | - | 2 - 3 | 386 | 67 |
AK160385.1 | - | 1 - 2 | 4399 | 67 | |
BC020368.1 | - | 1 - 2 | 688 | 67 | |
BC132777.1 | - | 1 - 2 | 347 | 67 | |
BC136512.1 | - | 1 - 2 | 345 | 67 |